Order Entry
ContactUsLinkComponent
MEDIA F-12K W/GLN 500ML PK6
Corning® F-12K Nutrient Mixture (Kaighn's Mod.) with L-glutamine, Corning
Catalog #: CA45000-354
Supplier:  Corning
CAS Number:  
MEDIA F-12K W/GLN 500ML PK6
Corning® F-12K Nutrient Mixture (Kaighn's Mod.) with L-glutamine, Corning
Catalog #: CA45000-354
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    F-12K Nutrient Mixture (Kaighn's Mod.) with L-glutamine
  • Storage temperature:
    Refrigerate
  • Cat. no.:
    CA45000-354
  • Supplier No.:
    10-025-CV

Specifications

About this item

A modification of Ham's F-12, F-12K contains higher concentrations of amino acids, sodium pyruvate and other components than the original

  • Low endotoxin: >0.25EU/mL
  • Mycoplasma tested
  • Formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment

It is formulated to support the growth of cells including differentiated rat and chicken cell types and primary human hepatocytes.

Customers Who Bought This Also Bought