Order Entry
ContactUsLinkComponent
DMEM LOGLU GLN + PYR 6X500ML
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Catalog #: CA45000-332
Supplier:  Corning
CAS Number:  
DMEM LOGLU GLN + PYR 6X500ML
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Catalog #: CA45000-332
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Corning® DMEM [+] 1 g/L glucose, L-glutamine, sodium pyruvate
  • Storage temperature:
    2...8 °C
  • Shelf Life:
    18 months
  • Supplier No.:
    10-014-CV

Specifications

About this item

Dulbecco's Modified Eagle's Medium (DMEM) is a modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME

  • Low endotoxin - <0.25 EU/mL
  • Mycoplasma tested
  • Formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment

The original formulation was used to culture embryonic mouse cells and has since been modified to support a variety of normal and transformed cells, such as primary cultures of mouse and chicken cells.

Customers Who Bought This Also Bought