Order Entry
ContactUsLinkComponent
Isotype: ; Applications: Fc; Reactivity:
Human TruStain FcX™, Fc Receptor Blocking Solution, BioLegend
Catalog #: CA422302-BL
Supplier:  Biolegend
CAS Number:  
Human TruStain FcX™, Fc Receptor Blocking Solution, BioLegend
Catalog #: CA422302-BL
Supplier:  Biolegend
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Human TruStain FcX™ Blocking Buffer
  • Cat. no.:
    CA422302-BL
  • Supplier No.:
    422302

Specifications

About this item

Human Fc receptors (FcRs) are expressed on a variety of cells, such as monocytes, granulocytes, B cells and dendritic cells

The cells with FcR expression sometimes give false positive or false negative results of immunofluorescent staining due to the FcRs-mediated Ig Fc binding. Human TruStain FcX™ is specially formulated for blocking the FcR-involved unwanted staining without interfering with antibody-mediated specific staining of human cells. This buffer contains specialized human IgG. It is not recommended to be used for staining human IgG. Handle as biohazard agent. Soluble.

Customers Who Bought This Also Bought