To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Control
- Antigen name:IgM, k Isotype Ctrl
- Clonality:Monoclonal
- Clone:MM-30
- Host:Mouse
- Isotype:IgM kappa
- Reactivity:Mouse
- Antigen symbol:IgM, k Isotype Ctrl
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:50 µg
- Western blot:Yes
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:The immunoglobulin solution should be stored undiluted between 2 °C and 8 °C
- Concentration:0.50 mg/mL
- Shipping temperature:Ambient RT
- Immunogen:Trinitrophenol + KLH
- Purification:The antibody is at >85% purity.
- Cat. no.:CA401601-BL
- Supplier No.:401601
Specifications
About this item
Purified Mouse IgM, κ Isotype Ctrl [MM-30]; Isotype: Mouse IgM, κ; Reactivity: Mouse; Apps: FC, ICFC, ICC, IF; Size: 500 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control, and the purity is greater than 85% by SDS-PAGE. Use at concentrations comparable to those of the specific antibody of interest.
Type: Control
Antigen: IgM, k Isotype Ctrl
Clonality: Monoclonal
Clone: MM-30
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgM, κ
Reactivity: Mouse