To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Control
- Antigen name:IgG1, k Isotype Ctrl
- Clonality:Monoclonal
- Clone:RTK2071
- Host:Rat
- Isotype:IgG1 kappa
- Reactivity:Rat
- Antigen symbol:IgG1, k Isotype Ctrl
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:500 µg
- Western blot:Yes
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:The IgG1 kappa isotype control solution should be stored undiluted between 2 °C and 8 °C.
- Concentration:0.50 mg/mL
- Shipping temperature:Ambient RT
- Immunogen:Trinitrophenol + KLH
- Purification:The antibody was purified by affinity chromatography.
- Cat. no.:CA400402-BL
- Supplier No.:400402
Specifications
About this item
Purified Rat IgG1, κ Isotype Ctrl [RTK2071]; Isotype: Rat IgG1, κ; Reactivity: Rat; Apps: FC, ICFC, ICC, IF, IHC, IP, WB; Size: 500 μg
Each lot of this rat IgG1, κ isotype control antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control, and the purity is greater than 95% by SDS-PAGE. Use at concentrations comparable to those of the specific antibody of interest. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.
Type: Control
Antigen: IgG1, k Isotype Ctrl
Clonality: Monoclonal
Clone: RTK2071
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Rat