To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD107a
- Clonality:Monoclonal
- Clone:H4A3
- Gene ID:3916
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Baboon,African green monkey,Cynomolgus monkey,Rhesus monkey,Pigtailed macaque,Chimpanzee,Human
- Antigen symbol:CD107a
- Conjugation:Alexa Fluor® 647
- Flow cytometry:Yes
- ImmunoFluorescence:Yes
- Size:25 tests
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Antigen synonyms:LAMP1|LGP-120
- Storage temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:Ambient RT
- Immunogen:Human adult adherent peripheral blood cells
- Purification:The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 647 under optimal conditions.
- Cat. no.:CA328611-BL
- Supplier No.:328611
Specifications
About this item
Alexa Fluor® 647 anti-human CD107a (LAMP-1) [H4A3]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus; Apps: FC, IF; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD107a
Clonality: Monoclonal
Clone: H4A3
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Chimpanzee;Cynomolgus;Pigtailed Macaque;Rhesus