Order Entry
ContactUsLinkComponent
MINIMATE TFF 1000K OMEGA
Minimate™ Tangential Flow Filtration Capsules, Cytiva (Formerly Pall Lab)
Catalog #: CA29301-924
Supplier:  Cytiva
MINIMATE TFF 1000K OMEGA
Minimate™ Tangential Flow Filtration Capsules, Cytiva (Formerly Pall Lab)
Catalog #: CA29301-924
Supplier:  Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Minimate capsule with Omega membrane
  • Cat. no.:
    CA29301-924
  • MWCO:
    1000K
  • Supplier No.:
    OA990C12

Specifications

About this item

The disposable Minimate™ TFF capsules, with Omega™ membrane, can perform concentration processes desalting or buffer exchange on the same system with minimal user intervention, and achieves high concentration factors in a single step due to the low hold-up volume of the devices.

  • Scaleable
  • Works with a variety of positive pressure laboratory pumps
  • High product recovery
  • 100% integrity tested

The capsules will process samples of up to 1 litre, or efficiently concentrate samples as little as 5 ml.

Delivery: Supplied with all fittings and tubing to simplify installation into filtration setups. A Certificate of Quality is included with each capsule.
Customers Who Bought This Also Bought