Specifications
- Dimensions:3.3 m
- Description:FluoroTrans® W PVDF membrane, roll, pore size 0.2 µm
- Cat. no.:CA29301-856
- Supplier No.:BSP0161
Specifications
About this item
Naturally hydrophobic PVDF, ideal for a wide variety of protein-analysis applications. The family of FluroTrans® media are white, microporous solid phase supports that bind proteins tenaciously via hydrophobic interactions. FluoroTrans® membrane is optimised for N-terminal protein sequencing. FluoroTrans® W membranes are optimised for Western transfer applications. They allow for sensitive protein detection with low background and very low protein burn-through. Immobilised proteins can be visually detected with Coomassie® Blue, Amido black, Ponceau S, and colloidal gold.
Filter membranes are resistant to a wide range of chemical solvents (i.e., acetone, DMSO, dimethyl formamide) and will not shrink during destaining with methanol. Resistance to trifluoroacetic acid and triethylamine allows direct insertion of FluoroTrans® membrane materials into amino acid analyzers and gas phase protein sequencing equipment.
- High protein binding capacity - typically absorbs 50% more protein than nylon or nitrocellulose
- High tensile strength, so won't tear, crack or curl during handling, allowing easy removal of target bands for protein sequencing applications
- High sensitivity for small peptides
Naturally hydrophobic PVDF, ideal for a wide variety of protein-analysis applications. The family of FluroTrans® media are white, microporous solid phase supports that bind proteins tenaciously via hydrophobic interactions. FluoroTrans® membrane is optimised for N-terminal protein sequencing. FluoroTrans® W membranes are optimised for Western transfer applications. They allow for sensitive protein detection with low background and very low protein burn-through. Immobilised proteins can be visually detected with Coomassie® Blue, Amido black, Ponceau S, and colloidal gold.
Pore size: 0,2 µm
Pore size: 0,2 µm
Typical thickness: 127 µm
Typical thickness: 127 µm