Order Entry
ContactUsLinkComponent
Unlined Viton Over Butyl Gloves
Viton® Unsupported Gloves, Showa
Catalog #: CA28888-328
Supplier:  Showa-Best Glove
Unlined Viton Over Butyl Gloves
Viton® Unsupported Gloves, Showa
Catalog #: CA28888-328
Supplier:  Showa-Best Glove

Specifications

  • Ambidextrous/Hand-specific:
    Ambidextrous
  • Glove Color:
    Black
  • Glove size:
    S
  • Length:
    30.5 cm (12")
  • Thickness:
    12 mil
  • Cat. no.:
    CA28888-328
  • Supplier No.:
    892-7

Specifications

About this item

Best® Viton® Fluoroelastomer rubber provides superior resistance to highly corrosive acids.

  • Excellent for handling aliphatic and aromatic hydrocarbons
  • Chemical resistant glove

This synthetic rubber provides the highest permeation resistance to high molecular aromatic hydrocarbons such as Benzene, Toluene, Xylene and Polychlorinated Biphenyls (PCBs) of any protective material used to make gloves.

These are used in military applications, acids and caustics, chlorinated solvents fuels etc.

Viton gloves provide the ultimate combination of protection and dexterity.

Customers Who Bought This Also Bought