Whatman™ Nylon Membrane Circles
Catalog #: CA28159-720
Supplier: Cytiva
Specifications
- Ø:13 mm
- Pore size:0.2 µm
- Description:Nylon membrane circle
- Material:Nylon
- Thickness:150 - 187 µm
- Max. operating temperature:135 °C
- Application:Filtration of aqueous and organic mobile phases
Vacuum degassing
Filtration of tissue culture media, microbiological media, buffers, and solutions - Type:Hydrophilic
- Water Flowrate:>50 ml/min (at 5 psi)
- Supplier No.:7402-001
Specifications
About this item
High-quality nylon membranes are suitable for filtering aqueous solutions and most organic solvents. The membranes are suitable for use with a wide range of biological preparations and can be used where other membranes are unsuitable or difficult to use.
- Flexible, durable and tear resistant
- Can be autoclaved at 135 °C
Nylon membranes are hydrophilic, eliminating the need for wetting agents that could be extracted when filtering aqueous solutions.
Customers Who Bought This Also Bought