Order Entry
ContactUsLinkComponent
Versapor* 1200 Membrane Disc Filter
Versapor® Membrane Disc Filters, Cytiva (Formerly Pall Lab)
Catalog #: CA28149-718
Supplier:  Cytiva
Versapor* 1200 Membrane Disc Filter
Versapor® Membrane Disc Filters, Cytiva (Formerly Pall Lab)
Catalog #: CA28149-718
Supplier:  Cytiva

Specifications

  • Ø:
    293 mm
  • Pore size:
    1.20 µm
  • Cat. no.:
    CA28149-718
  • Supplier No.:
    66397

Specifications

About this item

Ideal for prefiltration of difficult-to-filter solutions and serum.

Filter membrane is made of acrylic copolymer on a nonwoven support. Strong, flexible, and nonshedding, both dry and wet. Delivers high flow rates at low differential pressures. Can be used as a prefilter to replace glass fiber filters. UV or gamma sterilizable.

  • For prefiltration
  • Excellent retention
  • Acrylic copolymer

Ideal for prefiltration of difficult-to-filter solutions and serum.

Designed to be used in applications requiring excellent retention properties combined with tensile strengths in excess of 200 bar (3000 psi).

Filter membrane is made of acrylic copolymer on a nonwoven support. Strong, flexible, and nonshedding, both dry and wet. Delivers high flow rates at low differential pressures. Can be used as a prefilter to replace glass fiber filters. UV or gamma sterilizable.

Ordering information: For additional diameters, please contact your Avantor sales representative.
Customers Who Bought This Also Bought