Order Entry
ContactUsLinkComponent
MAG FUNNEL HSG 150ML PPS
Accessories for Magnetic Filter Funnels, 47 mm, Cytiva (Formerly Pall Lab)
Catalog #: CA28143-526
Supplier:  Cytiva
MAG FUNNEL HSG 150ML PPS
Accessories for Magnetic Filter Funnels, 47 mm, Cytiva (Formerly Pall Lab)
Catalog #: CA28143-526
Supplier:  Cytiva

Specifications

  • Description:
    Funnel cylinder, 150 ml, for use with 47 mm Ø filter
  • Cat. no.:
    CA28143-526
  • Supplier No.:
    4248

Specifications

About this item

Customers Who Bought This Also Bought