Syringe filters, Acrodisc®, Supor® membrane, Cytiva (Formerly Pall Lab)
Catalog #: CA28143-046
Supplier: Cytiva
Specifications
- Ø:32 mm
- Pore size:1.20/0.45 µm
- Sterility:Non sterile
- Membrane:Supor
- Supplier No.:4661
Specifications
About this item
Acrodisc™ syringe filters with Supor™ membrane deliver high flow rates and low protein binding.
- Superior flow rates and higher throughputs than competitive devices
- Low protein binding to minimize sample loss
- Pre-sterilized products are sterilized by gamma irradiation to eliminate potential contamination by EtO residuals
- A range of sizes (13 to 37 mm) accommodates sample volumes from < 10 to 150 ml
- Acrodisc PF and Serum Acrodisc syringe filters feature built-in prefilter for increased throughput of difficult-to-filter liquids
- Reduces mycoplasma with the use of 0,1 μm pore size
Supor™ hydrophilic polyethersulfone (PES) membrane: PP housing (except 25 and 32 mm: Acrylic and Acrodisc Serum: ABS)
Applications:
0.1 and 0.2 μm pore sizes provide sterilization of small volumes of buffers, culture media, and additives.
Acrodisc PF and Serum Acrodisc syringe filters are ideal for clarification/sterilization of viscous or particulate-laden solutions.
Use larger pore size filters for prefiltration and particulate removal.
Certifications: Passes United States Pharmacopeia (USP) Biological Reactivity Test, In Vivo <88>.
Customers Who Bought This Also Bought