Acrodisc® Units with Mustang® E Membrane, Cytiva (Formerly Pall Lab)
Catalog #: CA28140-521
Supplier: Cytiva
Specifications
- Ø:25 mm
- Pore size:0.20 µm
- Sterility:Gamma irradiated
- Cat. no.:CA28140-521
- Supplier No.:MSTG25E3
Specifications
About this item
Disposible, single-use units remove endotoxins in water, buffers, neutral sugars, and certain biological solutions.
- Disposable
- Single-use
- Gamma irradiated
Disposible, single-use units remove endotoxins in water, buffers, neutral sugars, and certain biological solutions.
Flow rate of 1 to 4 ml/min. (varying with type of solution, concentration of proteins, and other components).
Bed volume: 0.12 ml
Differential pressure: <0.07 bar (7 kPa, 1 psi) @ 1 ml/min.
Max. operating pressure: 2.1 bar (210 kPa, 30 psi) at 60 ˚C (140 ˚F) or 5.5 bar (550 kPa, 80 psi) at 21 to 24 ˚C (70 to 75˚F).
Customers Who Bought This Also Bought