Order Entry
ContactUsLinkComponent
SYRINGE FLTR 45UM PVDF PK1500
Whatman™ GD/X Syringe Filters, Prefilter, Non-Sterile, Whatman products (Cytiva)
Catalog #: CA28138-198
Supplier:  Cytiva
SYRINGE FLTR  45UM PVDF PK1500
Whatman™ GD/X Syringe Filters, Prefilter, Non-Sterile, Whatman products (Cytiva)
Catalog #: CA28138-198
Supplier:  Cytiva

Specifications

  • Ø:
    25 mm
  • Pore size:
    0.45 µm
  • Sterility:
    Non sterile
  • Volume:
    >10 ml
  • Max. operating pressure:
    5.2 bar (75 psi)
  • Dimensions:
    21.6×29.8 mm
  • Pre-filter:
    100% Borosilicate Glass Fiber
    GMF 150 10 µm: 1 µm
    GF/F 0.7 µm
  • Autoclavable:
    Autoclave at 121 °C (131 °C max) at 1 bar (15 psi) for 20 minutes
  • Filtration Area:
    4.6 cm²
  • Membrane:
    PVDF
  • Supplier No.:
    6873-2504

Specifications

About this item

Whatman GD/X Syringe Filters are high-viscosity filters that incorporate a prefilter to enable filtration of viscous, dense or otherwise hard-to-filter material with high solids content as in environmental and food analysis samples.

  • Recyclable after use

Whatman GD/X Syringe Filters are high-viscosity filters that incorporate a prefilter to enable filtration of viscous, dense or otherwise hard-to-filter material with high solids content as in environmental and food analysis samples.

Non sterile Whatman GD/X Syringe Filters combine a polypropylene housing, high-efficiency Whatman GMF 150 and GF/F pre-filter media, and one of the following membrane materials:

- Polyethersulfone (PES) for serum, plasma and tissue culture
- Polyvinylidene difluoride (PVDF) for HPLC
- Cellulose acetate for biological solutions
- Borosilicate glass microfiber

The prefilter contains a high-efficiency borosilicate glass filtration stack that will retain fine particles down to 0.7 μm, allowing difficult sample filtration with less hand force. It reduces the need for multiple syringe filters by processing three to seven times more sample volume than syringe filters without prefilters. The filters can be autoclaved.

Cytiva also offers Sterile Whatman GD/X syringe filters.

Customers Who Bought This Also Bought