ContactUsLinkComponent
TEST STRIP CHLORINE BOTTLE 50 STRIPS
Free and Total Chlorine Test Strips, Hach
Catalog #: CA27450-50
Supplier:  Hach
CAS Number:  
Free and Total Chlorine Test Strips, Hach
Catalog #: CA27450-50
Supplier:  Hach
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Range:
    0 - 10 mg/L
  • Supplier No.:
    27450-50
  • No. of Tests:
    50
Specifications
About this item
The most widely used disinfectant for drinking water, chlorine is also important for sanitizing swimming pools, cooling towers, other industrial equipment, and in the treatment of municipal wastewater. Its measurement and control are vital for both safety and economic reasons. Chlorine exists in water as free chlorine (hypochlorite) or combined chlorine (chloramines).
With Hach Water Quality Test Strips, technicians in the field can test many samples in only a few minutes and make immediate evaluations on-site. No measuring, set-up, clean-up, or chemical handling are necessary. Hach test strips are also used in laboratories all over the world for pre-test screening-to detect the presence of materials that might interfere with lab testing.
With Hach Water Quality Test Strips, technicians in the field can test many samples in only a few minutes and make immediate evaluations on-site
Accurate and easy to use - just dip and read. Great for quick, on-site evaluations of free and total chlorine levels.
No measuring, set-up, clean-up, or chemical handling are necessary. Hach test strips are also used in laboratories all over the world for pre-test screening-to detect the presence of materials that might interfere with lab testing.
Customers Who Bought This Also Bought