Specifications
- Dimensions:90 x 15 mm
- Description:Petri Dish, Sliding Version with 3 Vents and ISO Mark
- Material:Polystyrene
- Cat. no.:CA25389-330
Specifications
About this item
These semi-stacking Petri dishes are available with a semi-stacking ring for easy transit during automatic filling or in a sliding version with a totally smooth lid.
- Distortion-free viewing
- Versatile and suitable for different makes and models of pouring equipment
The controlled bottom flatness ensures even distribution of media and organisms. The semi-stacking ring has been specifically designed to make the dish versatile and suitable for different makes and models of pouring equipment.