Specifications
- Ø:60 mm
- Height:15 mm
- Cat. no.:CA25382-687
- Packaging:20/sleeve
- For Use With:Cell culture
- Supplier No.:353802
Specifications
About this item
PS, sterile, with lid.
- Unique, nitrogen-containing tissue culture surface chemistry improves attachment, spreading and growth for many primary cells or cell lines
- Optically clear and no special storage required
- Convenient, peel open, medical-style packaging
The gases used to manufacture Corning® Primaria™ contain both oxygen and ammonia, resulting in the incorporation in the surface of a variety of nitrogen-containing functional groups in addition to the negatively charged oxygen containing groups found on traditional tissue culture treated surfaces. The incorporation of nitrogen containing cations has been correlated to attachment and spreading of primary endothelial cells in a clonal cell-growth assay. The complex surface on Corning® Primaria™ products is homogeneous and stable and is used to improve attachment and differentiation of a variety of cell types. The surface chemistry of Corning® Primaria™ products is confirmed by Electron Scanning for Chemical Analysis (ESCA).