Nunclon™ Delta 96-Well MicroWell™ Plates, Sterile, Thermo Scientific
Catalog #: CA25382-342
Supplier: Thermo Fisher Scientific
Specifications
- Well Area:0.33 cm2
- Well Volume:0.4 mL
- Color:Clear
- Working volume:50 - 250 µl
- Packaging:1/Sleeve
- Well Shape:Flat-Bottom
- Supplier No.:167008
Specifications
About this item
These 96-well, polystyrene plates are ideal for cell culture, cloning, viral titration, and cell fusion
The Nunclon™Δ treated surface ensures optimal cell growth and attachment. Plates fit standard automated handling equipment. Plate edges and lids are designed to minimize evaporation. Lids are shaped to facilitate stacking. Raised well rims reduce cross-contamination. White plates provide maximum reflection, minimum autofluorescence and autoluminescence. Black plates provide minimum background and back light scatter in fluorescence assays. Plates also feature alphanumeric well identification and excellent optical quality. Radiation sterilized.
Ordering information: For lids and sealing tapes, see the Nunc® Microplate Lids, Thermo Scientific series.
Customers Who Bought This Also Bought