Order Entry
ContactUsLinkComponent
Nunclon* Delta Dishes, Polystyrene, Sterile
Nunc™ Cell Culture/Petri Dishes, Thermo Scientific
Catalog #: CA25382-334
Nunclon* Delta Dishes, Polystyrene, Sterile
Nunc™ Cell Culture/Petri Dishes, Thermo Scientific
Catalog #: CA25382-334

Specifications

  • Ø:
    35 mm
  • Description:
    Nunclon™Δ Treated
  • Height:
    10 mm
  • Growth Area:
    8.8 cm²
  • Diameter ×H:
    35×10 mm
  • Supplier No.:
    153066

Specifications

About this item

Perform a variety of applications across a broad range of sizes and styles to suit a variety of experimental needs with Thermo Scientific™ Nunc™ Cell Culture/Petri Dishes

These cell culture/petri dishes are made of polystyrene.

These dishes receive the Thermo Scientific™ Nunclon™ Delta surface treatment, a proprietary surface modification that promotes maximum adhesion for a broad range of cell types.

Ordering information: Quality certificates are included in each case.
Customers Who Bought This Also Bought