Order Entry
ContactUsLinkComponent
BD Falcon* Organ Culture Dish
Falcon® Organ Culture Dish, Sterile, Corning
Catalog #: CA25381-141
Supplier:  Corning
BD Falcon* Organ Culture Dish
Falcon® Organ Culture Dish, Sterile, Corning
Catalog #: CA25381-141
Supplier:  Corning

Specifications

  • Ø:
    60 mm
  • Dimensions:
    54.84 O.D. x 13.56H mm Actual
  • Description:
    Organ Culture Dish, Center Well Area 2.89 cm2
  • Height:
    15 mm
  • Cat. no.:
    CA25381-141
  • Supplier No.:
    353037

Specifications

About this item

Designed for tissue and organ culture techniques

Polystyrene dish is tissue culture treated by vacuum-gas plasma. Sterilized by gamma irradiation. Nonpyrogenic.

Ordering information: Packaged 20 per sleeve.
Customers Who Bought This Also Bought