ContactUsLinkComponent
Square Petri Dish with 13mm Grid
Square Polystyrene Disposable, Sterile Petri Dishes
Catalog #: CA25378-115
Supplier:  Simport Scientific
Square Petri Dish with 13mm Grid
Square Polystyrene Disposable, Sterile Petri Dishes
Catalog #: CA25378-115
Supplier:  Simport Scientific
Specifications
  • Description:
    Square Polystyrene Disposable, Sterile Petri Dishes
  • Form:
    Liquid
  • Cat. no.:
    CA25378-115
  • Supplier No.:
    D210-16
Specifications
About this item
Petri dishes having a raised square grid on the bottom part. The grid consists of 36 small squares; each square is identified to facilitate special type counting or location of specimens. Supplied with top and bottom in sterile shelf packages. Size: 100 x 15mm.
Customers Who Bought This Also Bought