Order Entry
ContactUsLinkComponent
Sterile Petri Dish
Falcon® Petri Dishes, Tight-Fit Lid, Sterile
Catalog #: CA25373-041
Supplier:  Corning
Sterile Petri Dish
Falcon® Petri Dishes, Tight-Fit Lid, Sterile
Catalog #: CA25373-041
Supplier:  Corning

Specifications

  • Ø:
    35 mm
  • Height:
    10 mm
  • Packaging:
    20/Sleeve
  • Diameter ×H:
    35 x 10 mm
  • Supplier No.:
    351008

Specifications

About this item

Dishes are constructed with crystal-grade polystyrene and a flat bottom surface for distortion-free optics

  • Flat, distortion free optics
  • Stacking rings allow for easier stacking and handling
  • Durable construction for stable dish manipulation

Small diameter saves media and space. Easily sealed with tape for collecting and transporting specimens. Polystyrene. Sterilized by gamma irradiation. Packed 20 per bag.

Customers Who Bought This Also Bought