Falcon® Petri Dishes, 50×9 mm, Tight-Fit Lid, Sterile, Corning
Catalog #: CA25369-022
Supplier: Corning
Specifications
- Description:Falcon* Petri Dishes
- Size:50 x 9 mm
- Cat. no.:CA25369-022
- Supplier No.:351006
Specifications
About this item
PS. Manufactured in accordance with the current FDA Quality System.
- Flat, distortion free optics
- Stacking rings allow for easier stacking and handling
- Durable construction for stable dish manipulation
Small diameter saves media and space. Easily sealed with tape for collecting and transporting specimens. Polystyrene. Sterilized by gamma irradiation. Packed 20 per bag.
Customers Who Bought This Also Bought