Order Entry
ContactUsLinkComponent
BOD SEED PK50
BOD Seed Inoculum, POLYSEED, Hach
Catalog #: CA24712-00
Supplier:  Hach
CAS Number:  
BOD SEED PK50
BOD Seed Inoculum, POLYSEED, Hach
Catalog #: CA24712-00
Supplier:  Hach
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    BOD Seed Inoculum, POLYSEED
  • Cat. no.:
    CA24712-00
  • Supplier No.:
    2918700

Specifications

About this item

BOD Seed Inoculum for use in Biochemical Oxygen Demand (BOD5) test procedures. A blend of specialized microbial cultures in an easy-to-use capsule. Designed to provide a uniform standard for the degradation of both industrial and municipal waste in BOD5 analyses.

  • Consitent seed source: same bacteria, same type of bacteria, same amount of bacteria blend
  • Known bacterial content is free from pathogens
  • Save and convenient pre-measured dry product in easy to use capsules

Polyseed®-NX CBOD contains an additive for the inhibition of ammonia nitrogen in a wastewater sample. Additive amount is comparable to that stated in Standard Methods as necessary for sufficient inhibition.

Two years shelf life

Application: Dilution water preparation. Method number: 8043

Ordering information: Package of 50 capsules.
Customers Who Bought This Also Bought