Order Entry
ContactUsLinkComponent
SAMPLE CELL W/CAP 36X60MM PK6
Sample Cells, Round Glass, Hach
Catalog #: CA24276-06
Supplier:  Hach
SAMPLE CELL W/CAP 36X60MM PK6
Sample Cells, Round Glass, Hach
Catalog #: CA24276-06
Supplier:  Hach

Specifications

  • Capacity:
    10 ml
  • Ø:
    1"
  • Description:
    Round Glass Sample Cells
  • Cat. no.:
    CA24276-06
  • Supplier No.:
    24276-06

Specifications

About this item

Round, glass sample cells for use with Hach Spectrophotometers

Cells come in pack of six with caps.

Customers Who Bought This Also Bought