ContactUsLinkComponent
DPD COMPOUND FREE + TOTAL 24GM
CL17 DPD Indicator, Hach
Catalog #: CA22972-55
Supplier:  Hach
CAS Number:  
DPD COMPOUND FREE + TOTAL 24GM
CL17 DPD Indicator, Hach
Catalog #: CA22972-55
Supplier:  Hach
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Chemical method:
    DPD (N,N-diethyl-p-phenylenediamine)
  • Packaging:
    Jar
  • Size:
    24 g
  • Cat. no.:
    CA22972-55
  • Supplier No.:
    22972-55
  • No. of Tests:
    450
Specifications
About this item
DPD is an indicator that is used to measure either Free Chlorine or Total Chlorine with the CL17 Analyzer. Depending upon whether Free Chlorine or Total Chlorine is being measured, the DPD indicator is added to Free Chlorine or Total Chlorine Indicator Solution and used with Free or Total Buffer Solution by the CL17 Analyzer. *This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
  • Quality, Reliability
  • Simple, Predictable Maintenance
  • Reagent Delivery Program
Customers Who Bought This Also Bought