To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Chemical method:DPD (N,N-diethyl-p-phenylenediamine)
- Description:DPD Total Chlorine Reagent, SwifTest™ Dispenser Refill Vial
- Size:10 mL Sample
- Method:8167 - DPD
- Compliance:EPA - Yes
- Instrument:All except DR/3000, DR/2000
- Cat. no.:CA21056-60
- Range:0.02 - 2.00 mg/l Cl₂
- Supplier No.:21056-60
- No. of Tests:250
Specifications
About this item
For total chlorine determination by the DPD Total Chlorine method.
-Fast, easy reagent addition
-Just squeeze to add powder
- Fast, easy reagent addition
- Just squeeze to add powder
For use with Hach SwifTest DPD dispenser. USEPA accepted for reporting for drinking and wastewater analysis using Hach Method 8167. Range: 0.02 - 2.00 mg/L Cl2.