J&W HP-1 Columns, Agilent Technologies
Catalog #: CA19091Z-215
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Column Format:Capillary
- Film thickness:1.05 µm
- I.D.:0.32 mm
- Length:50 m
- Description:GC column, 7" cage
- Polarity:Low-polarity stationary phase
- Column Hardware:Fused silica
- Cat. no.:CA19091Z-215
- Phase:HP-1
- Supplier No.:19091Z-215
Specifications
About this item
A nonpolar 100% polydimethylsiloxane, general-purpose, industry-standard column for the analysis of hydrocarbons, pesticides, PCBs, and sulfur compounds.
- 100% Dimethylpolysiloxane
- Non-polar
- Excellent general-purpose, industry-standard columns
- Superior performance with low molecular weight alcohols (< C5)
- Bonded, crosslinked, and solvent rinsable
- Wide range of column dimensions available
- Equivalent to USP phase G2
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
- Similar phases: SPB-1, Rtx-1, BP-1, OV-1, OV-101, 007-1(MS), SP-2100, SE-30, ZB-1, AT-1, MDN-1, ZB-1
Agilent J&W HP-1 is a general purpose 100% Dimethylpolysiloxane. Widely considered as an 'industry standard' column, it is an excellent choice for a range of applications that include analysis of hydrocarbons, pesticides, low molecular weight alcohols, polychlorinated biphenyls (PCBs), and sulfur compounds.
Customers Who Bought This Also Bought