ContactUsLinkComponent
CIP 100* Detergent System
CIP 100® Detergent System, Steris
Catalog #: CA16200-114
Supplier:  Steris
CIP 100® Detergent System, Steris
Catalog #: CA16200-114
Supplier:  Steris
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Pail
  • Size:
    18.9 L (5 gal.)
  • Supplier No.:
    1D1005
Specifications
About this item
CIP 100 alkaline detergent is ideal for clean-in-place (CIP) and clean-out-of-place (COP) applications.
  • Low foaming, formulated alkaline cleaner contains biodegradable surfactants
  • Extremely free-rinsing, penetrates, dissolves and removes a broad spectrum of soils using multiple cleaning mechanisms
  • Both specific and non-specific methods are available for detecting cleaning agent residues
High-performance, phosphate-free, alkaline detergent system is for use in manual, immersion, and recirculating spray equipment
A proprietary blend of potassium hydroxide, surfactants, chelants and other performance-enhancing ingredients provide multiple cleaning mechanisms. The low foaming product removes a wide range of process residues, from fertmentation by-products to silicone-based emulsions and lubricants.
It is specially formulated to meet the unique process cleaning demands found in the pharmaceutical, biotechnology, cosmetic, dietary supplement and other industries requiring an effective validatable cleaner.
Certifications: Manufactured in a cGMP compliant, FDA registered and ISO 9001/13485 certified facility
Customers Who Bought This Also Bought