Order Entry
ContactUsLinkComponent
Disposable 4.5 oz. Sample Containers, Polypropylene, Sterile
Falcon® Sample Containers, Polypropylene, Sterile, Corning
Catalog #: CA15704-051
Supplier:  Corning
Disposable 4.5 oz. Sample Containers, Polypropylene, Sterile
Falcon® Sample Containers, Polypropylene, Sterile, Corning
Catalog #: CA15704-051
Supplier:  Corning
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Capacity:
    110 ml (3.7 oz.)
  • Description:
    Containers with Lid
  • Packaging:
    Individually Wrapped
  • Supplier No.:
    354013

Specifications

About this item

PP, sterile, with or without PE lid.

Polyethylene lid is frosted for easy marking, and fits both container sizes.

  • Molded-in graduations for easy measurement
  • Inert and chemically resistant to commonly used laboratory reagents at room temperature

PP, sterile, with or without PE lid.

Convenient, single-use containers for collection, transport and storage of a wide variety of specimens.

Polyethylene lid is frosted for easy marking, and fits both container sizes.

Customers Who Bought This Also Bought