Falcon® Cell Scrapers, Sterile, Corning
Catalog #: CA15621-010
Supplier: Corning
Specifications
- Overall Length:25 cm
- Supplier No.:353086
Specifications
About this item
PS handle, TPE blade. Designed to provide maximum accessibility to the growth surfaces of a variety of culture vessels.
- Cross-ribbed PS handle provides greater rigidity, to ensure better control while scraping cells
- Highly compliant TPE blade pivots to provide multiple angles to remove cells from the entire growth surface
Ordering information: Supplied individually wrapped in peel-open packages.
Customers Who Bought This Also Bought