Specifications
- Color:Lilac
- Cat. no.:CA14230-486
- Supplier No.:M485-10
Specifications
About this item
These disposable cassettes hold tissue specimens efficiently during the embedding process and in storage
These disposable cassettes hold tissue specimens efficiently during the embedding process and in storage
Molded from acetal polymer, they keep specimens submerged in liquid and are resistant to the chemical action of histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. Anterior writing area is angled at 45° for suitability with certain types of cassette labeling instruments. Colors are non-cytotoxic and nonmetallic