Order Entry
ContactUsLinkComponent
Blood Dilution Vials
Blood Dilution Vials, Simport Scientific
Catalog #: CA13915-951
Supplier:  Simport Scientific
Blood Dilution Vials
Blood Dilution Vials, Simport Scientific
Catalog #: CA13915-951
Supplier:  Simport Scientific

Specifications

  • Cap included:
    Yes
  • Capacity:
    25.00 ml
  • Cat. no.:
    CA13915-951
  • Packaging:
    Bulk Pack
  • Diameter ×H:
    35×56 mm (13/8×21/4")
  • Supplier No.:
    V130-1

Specifications

About this item

Made of polystyrene.

  • Optically clear parallel sides
  • Deionized
  • Destaticized

The snap cap is leakproof and easy to remove. Each lot is tested to ensure cleanliness. Available in specially designed vacuum formed trays or in bulk packaging.

Certifications: Guaranteed to meet or exceed O.E.M. specifications.
Customers Who Bought This Also Bought