Short-Term Detector Tubes, Draeger
Catalog #: CA13453-394
Supplier: Draeger
Specifications
- Gas or Vapor Tube:Oil
- Standard Measurement Range:0.1 - 1 mg/m³
- Supplier No.:6728371
Specifications
About this item
Short-Term detector tubes for use with the accuro® Gas Detector Pump and Aerotest Breathing Air Tester.
- Dräger Tubes are glass vials filled with a chemical reagent that reacts to a specific chemical or family of chemicals
- A variety of tubes are available for the measurement of numerous different gases and vapors
Ordering information: For additional tubes, contact your Avantor sales representative.
Customers Who Bought This Also Bought