DuPont™ ProClean® Frocks, Front Snap Closure
Catalog #: CA13452-504
Supplier: DuPont
Specifications
- Clothing Size:M
- Garment application:Laboratory
- Length:40"
- Color:White
- Cat. no.:CA13452-504
- Chest Size:241/2"
- Supplier No.:PC270SWHMD0B
Specifications
About this item
ProClean® offers improved non-hazardous light liquid splash protection.
- Serged seams
- Bound neck
- Set sleeve design
- Covered elastic wrists
- Front snap closure
ProClean® is made from a microporous composite fabric. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Covered elastic openings at wrists allows for a tighter fit. Bound neck ensures lower particle shedding. Frocks provide body-length jacket design with front snap closures for easy donning and doffing.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.
Customers Who Bought This Also Bought