Dupont™ ProShield® 60 Coveralls with Laydown Collar and Open Wrists
Catalog #: CA13452-250
Supplier: DuPont
Specifications
- Clothing Size:S
- Color:White
- Cat. no.:CA13452-250
- Chest Size:211/2"
- Inseam:261/2"
- Supplier No.:NG120SWHSM00
- Sleeve:311/4"
Specifications
About this item
ProShield® 60 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
- Laydown collar
- Open wrists and ankles
- Serged seams
ProShield® 60 garments are made with a microporous film that is laminated to a spundbonded polypropylene substrate. Applications include automotive refinishing, waste cleanup and remediation. Coveralls feature a laydown collar and open wrists and ankles.
Certifications: Thickness meets ASTM D751.
Customers Who Bought This Also Bought