Dupont™ ProShield® 60 Coveralls with Laydown Collar and Elastic Wrists
Catalog #: CA13451-006
Supplier: DuPont
Specifications
- Clothing Size:XL
- Color:White
- Cat. no.:CA13451-006
- Chest Size:27"
- Inseam:273/4"
- Supplier No.:NG125SWHXL00
- Sleeve:341/2"
Specifications
About this item
ProShield® 60 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
- Laydown collar
- Elastic wrists and ankles
- Serged seams
ProShield® 60 garments are made with a microporous film that is laminated to a spunbonded polypropylene substrate. Applications include automotive refinishing, waste cleanup and remediation. Coveralls feature a laydown collar and elastic opening at wrists and ankles for a tighter fit.
Certifications: Thickness meets ASTM D751.
Customers Who Bought This Also Bought