Short-Term Detector Tubes, Draeger
Catalog #: CA13450-568
Supplier: Draeger
Specifications
- Cat. no.:CA13450-568
- No. of Strokes:"1, 10"
- Gas or Vapor Tube:Hydrogen sulfide
- Standard Measurement Range:10 - 200 ppm, 1 - 20 ppm
- Supplier No.:8101831
Specifications
About this item
Short-Term detector tubes for use with the accuro® Gas Detector Pump and Aerotest Breathing Air Tester.
- Dräger Tubes are glass vials filled with a chemical reagent that reacts to a specific chemical or family of chemicals
- A variety of tubes are available for the measurement of numerous different gases and vapors
Ordering information: For additional tubes, contact your Avantor sales representative.
Customers Who Bought This Also Bought