Corning® BioCoat® Collagen I-Coated Flasks, Corning
Catalog #: CA12777-076
Supplier: Corning
Specifications
- Cap Type:Vented cap
- Capacity:250 ml
- Cat. no.:CA12777-076
- Packaging:5 per pack/50 per case
- Neck Style:Canted
- Growth Area:75 cm²
- Supplier No.:356485
Specifications
About this item
Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. In vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.
- Promotion of cell attachment and spreading
- Rapid expansion of cell populations
- Serum-free or reduced serum culture
- Cell adhesion assays
- Improving survival of primary cells in culture
- Storage: 4 to 30 °C under dry conditions
Customers Who Bought This Also Bought