Order Entry
ContactUsLinkComponent
Endothelial Micro Vascular Bulletkit
Endothelial Cell Growth Media Kits, Lonza
Catalog #: CA12002-220
CAS Number:  
Endothelial Micro Vascular Bulletkit
Endothelial Cell Growth Media Kits, Lonza
Catalog #: CA12002-220
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    EGM® -2 MV Microvascular Endothelial Cell Growth Medium-2 BulletKit®
  • Size:
    1 Kit, 500 ml
  • Cat. no.:
    CA12002-220
  • Application:
    Aortic and Pulmonary Artery Endothelial Cells
  • Supplier No.:
    CC-3202

Specifications

About this item

Clonetics™ offers a variety of different endothelial media kits.

They have varied formulations and are optimized to grow certain primary cell types and are guaranteed to perform when used to grow Clonetics™ Primary Cells.

Customers Who Bought This Also Bought