Order Entry
ContactUsLinkComponent
Pk One
ProKlenz® ONE Alkaline Detergent, STERIS®
Catalog #: CA11007-736
Supplier:  Steris
ProKlenz® ONE Alkaline Detergent, STERIS®
Catalog #: CA11007-736
Supplier:  Steris
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Capacity:
    18.93 L
  • Description:
    ProKlenz® ONE Alkaline Cleaner, Light Yellow, Odorless
  • Cat. no.:
    CA11007-736
  • pH:
    12.5
  • Specific Gravity:
    1.19
  • Supplier No.:
    142105EC

Specifications

About this item

ProKlenz® ONE High performance alkaline cleaner is ideal for use in clean-in-place (CIP), clean-out-of-place (COP) and manual applications.

  • NaOH-based detergent, environmentally friendly
  • Delivers effective microbial control in critical manufacturing processes with its disinfectant and virucidal properties
  • Manufactured with biodegradable ingredients to support harmonization of cleaning processes
  • An extensive support package is available to meet validation objectives

It is a proprietary blend of a concentrated alkalinity source, advanced surfactant system and other performance-enhancing ingredients provide multiple cleaning mechanisms

It is specially formulated to meet the unique cleaning demands found in the pharmaceutical, biotechnology, cosmetic, nutraceutical, and other industries requiring an effective validatable cleaner.

This low foaming product removes a wide range of process residues, from fermentation by-products to silicone-based emulsions and lubricants.

Certifications: Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility.
Customers Who Bought This Also Bought