Order Entry
ContactUsLinkComponent
KIMBERLY-CLARK SCOTT SCOTTFOLD HANDTOWELS
Scott® Scottfold* Towels, KIMBERLY-CLARK PROFESSIONAL®
Catalog #: CA11007-000
Supplier:  Kimberly-Clark
KIMBERLY-CLARK SCOTT SCOTTFOLD HANDTOWELS
Scott® Scottfold* Towels, KIMBERLY-CLARK PROFESSIONAL®
Catalog #: CA11007-000
Supplier:  Kimberly-Clark

Specifications

  • W×L:
    17.8 x 30.5 cm (7.8 x 12.4")
  • Color:
    White
  • Description:
    Scott Scottfold Towels
  • Cat. no.:
    CA11007-000
  • Packaging:
    175 Towels/25 Pack
  • Supplier No.:
    01960

Specifications

About this item

With quality and performance, the Scott Scottfold Towels can help reduce maintenance time with innovations like touchless, high-capacity systems.

Delivers one-at-a-time dispensing every time.

Certifications: FSC® and EcoLogo® certified meets EPA standards - Minimum of 40% post-consumer recycled fiber and total minimum of 50% recycled fiber.
Ordering information: Larger case size to reduce packaging waste.
Customers Who Bought This Also Bought