Order Entry
ContactUsLinkComponent
E.Z.N.A. FFPE ISO KIT 50 PRPS
E.Z.N.A.® FFPE DNA Kit, Omega
Catalog #: CA11006-984
Supplier:  Omega Bio-Tek
CAS Number:  
E.Z.N.A.® FFPE DNA Kit, Omega
Catalog #: CA11006-984
Supplier:  Omega Bio-Tek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    FFPE tissues
  • Target molecule:
    DNA
  • Description:
    E.Z.N.A.® FFPE DNA Kit
  • Size:
    50 Preps
  • Description:
    E.Z.N.A.® FFPE DNA Kit
  • Cat. no.:
    CA11006-984
  • Supplier No.:
    D3399-01

Specifications

About this item

The E.Z.N.A.® FFPE DNA Kit is designed for fast and easy purification of DNA from formalin-fixed, paraffin-embedded (FFPE) tissue sections

After adjusting the binding conditions with ethanol, the lysate is applied to the MicroElute DNA column to bind DNA. Cellular debris and proteins are effectively removed during the wash steps. High quality purified DNA is then eluted in elution buffer or water and is ready for applications such as PCR and next-generation sequencing.

Paraffin removal can be performed using a xylene and ethanol method or an efficient heat treatment. Samples are incubated in a specialized lysis buffer along with Proteinase K to reverse crosslinking, effectively releasing short and long DNA fragments.

Customers Who Bought This Also Bought