Order Entry
ContactUsLinkComponent
Polystyrene Antistatic Square Weighing Dishes 100mL
Antistatic Weigh Dishes, Simport Scientific
Catalog #: CA11006-614
Supplier:  Simport Scientific
Polystyrene Antistatic Square Weighing Dishes 100mL
Antistatic Weigh Dishes, Simport Scientific
Catalog #: CA11006-614
Supplier:  Simport Scientific

Specifications

  • Capacity:
    100 ml
  • Dimensions:
    78×78×25 mm
  • Description:
    Weighing Dishes
  • Cat. no.:
    CA11006-614
  • Packaging:
    250/Pack, 500/Case
  • Supplier No.:
    D250-2

Specifications

About this item

Weighing Dishes will resist diluted acids, aqueous solutions, alcohols and bases, ideal for many applications such as weighing, dispensing, or storing.

  • Safe, contaminant-free, biologically inert economical containers
  • For weighing liquid or powdered samples in the laboratory
  • Flat bottom ensures perfect stability on countertops

Dishes can also be used as quick freeze trays for sample material, discard trays for broken ampules, or mixing trays for small batches. Will withstand temperatures up to 80 °C.

Customers Who Bought This Also Bought