Order Entry
ContactUsLinkComponent
Phospholipids C Kit
Phospholipids C, Wako
Catalog #: CA11001-736
Supplier:  Wako Life Sciences
CAS Number:  
Phospholipids C, Wako
Catalog #: CA11001-736
Supplier:  Wako Life Sciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Phospholipids C
  • Size:
    8 x 50 mL, 2 x 10 mL (kit)
  • Storage temperature:
    2-10°C
  • Supplier No.:
    433-36201

Specifications

About this item

The Wako Phospholipids C is an in vitro assay for the quantitative determination of phospholipids in serum.

  • Phospholipid specific enzymatic assay
  • Measures Sphingomyelin, Lecithin, and Lysolecithin
  • Linear up to 1000 mg/dL

Method: Choline oxidase-DAOS

Phospholipids play an important function in the composition of cell membranes and in the emulsification and absorption of fat in the body. Serum phospholipids are formed in the liver, where they become bound to apolipoproteins, which makes them soluble in plasma. The Wako Phospholipids C assay is an enzymatic method utilizing choline oxidase in a reaction which can be measured photometrically. This assay has been shown not to be significantly influenced by coexisting substances such as ascorbic acid and bilirubin.

Composition
Buffer Solution: 8 x 50 mL
Color Reagent: 8 x for 50 mL
Standard Solution: 2 x 10 mL

Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought