KLEENGUARD® A10 Light Duty Apron, KIMBERLY-CLARK PROFESSIONAL®
Catalog #: CA10837-376
Supplier: Kimberly-Clark
Specifications
- Garment application:Industrial,General Purpose
- W×L:71.1×91.4 cm (28×36")
- Color:White
- Environmentally Preferable:Waste Reducing
- Cat. no.:CA10837-376
- Supplier No.:43744
Specifications
About this item
Disposable aprons provide light-duty protection against dirt, grime, and certain dry particulates in non-hazardous environments
Lightweight, polyethylene-coated, spunbound polypropylene fabric.
Customers Who Bought This Also Bought