KLEENGUARD® A10 Light Duty Shoe Covers, KIMBERLY-CLARK PROFESSIONAL®
Catalog #: CA10837-364
Supplier: Kimberly-Clark
Specifications
- Shoe size:Universal
- Color:Blue
- Cat. no.:CA10837-364
- Supplier No.:36811
Specifications
About this item
Shoe covers are designed to provide light duty protection in food processing, general maintenance, and manufacturing applications
- Blue colour
- Fully elasticized, construction ensures a secure fit
- Low skid feature
Covers are manufactured from a lightweight spunbond polypropylene material that provides great comfort throughout extended use. Provides the right combination of price and protection for non-hazardous environments.
Customers Who Bought This Also Bought