KLEENGUARD® A20 Breathable Particle Protection Shirts, KIMBERLY-CLARK PROFESSIONAL®
Catalog #: CA10837-282
Supplier: Kimberly-Clark
Specifications
- Anti-static:Yes
- Clothing Size:L
- Garment application:Medical,Industrial,General Purpose
- Closure type:Press studs / snaps
- Color:White
- Cat. no.:CA10837-282
- Supplier No.:36213
Specifications
About this item
Made from a patented Microforce™ Barrier SMS Fabric that offers barrier protection against dry particulates and light liquid sprays
The lightweight and breathable properties of the fabric make this shirt comfortable to wear, while the strength and abrasion-resistant properties extend the use of this product. NFPA 99 Compliant Antistatic Material. Five snap front closure.
Customers Who Bought This Also Bought