KLEENGUARD® A20 Breathable Particle Protection Frocks, Kimberly-Clark
Catalog #: CA10815-986
Supplier: Kimberly-Clark
Specifications
- Clothing Size:XXL
- Garment application:Laboratory
- Gender:Unisex
- Pockets:Without pockets
- Closure type:Press studs / snaps
- Color:White
- Fabric:SMMMS
- Cat. no.:CA10815-986
- Supplier No.:36265
Specifications
About this item
KLEENGUARD® A20 SMMMS frocks offer superior particle holdout coupled with unmatched comfort attributes, providing workers protection without sacrificing comfort against non-hazardous and hazardous dust
- Superior particle holdout coupled with unmatched comfort attributes
- 5 layers of stacked barrier protection (SMMMS) against hazardous particles
Lightweight and breathable. Serged seams, five-snap closure, elastic wrists, below knee length, and mandarin collar.
The A20 fabric has five layers of stacked barrier protection (SMMMS) against hazardous particles.
Certifications: NFPA 99 for Antistatic Materials. Eligible for recycling with KIMBERLY-CLARK PROFESSIONAL®
Customers Who Bought This Also Bought